CH6371380370 Chem Impex International, Inc.

Cyclo(-Gly-L-Glu)

  • MDL number MFCD00237613
  • Cas Number : 16364-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Cyclo(-Gly-Glu)
Storage temp. Store at =-4
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-gly-l-glu/5339?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-gly-l-glu/5339?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart