CH6371380375 Chem Impex International, Inc.

Cyclo(-Gly-Trp)

  • MDL number MFCD00083708
  • Cas Number : 7451-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-gly-trp/5344?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-gly-trp/5344?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart