CH6371380376 Chem Impex International, Inc.

Cyclo(Gly-L-Leu)

  • MDL number MFCD00076706
  • Cas Number : 5845-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Cyclo(-Leu-Gly)
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-gly-l-leu/5345?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-gly-l-leu/5345?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart