CH6371380378 Chem Impex International, Inc.

Cyclo(-L-Leu-L-Pro)

  • MDL number MFCD00237617
  • Cas Number : 2873-36-1
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-l-leu-l-pro/5347?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-l-leu-l-pro/5347?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart