CH6371380379 Chem Impex International, Inc.

Cyclo(L-Phe-L-Phe)

  • MDL number MFCD00190985
  • Cas Number : 2862-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Cyclo(-Phe-Phe)
Purity 99
Storage temp. Store at = -4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/cyclo-l-phe-l-phe/5350?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/cyclo-l-phe-l-phe/5350?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart