CH6371380387 Chem Impex International, Inc.

Deamino-histidine

  • MDL number MFCD00237415
  • Cas Number : 1074-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Imidazol-4-yl)propionic acid
Purity 98
Storage temp. Store at =-15
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/deamino-histidine/5360?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/deamino-histidine/5360?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart