CH6371380388 Chem Impex International, Inc.

Pyr-Gln-Ala-OH

  • MDL number MFCD00236971
  • Cas Number : 21477-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Eisenin
Storage temp. Store at =-4
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/pyr-gln-ala-oh/5364?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pyr-gln-ala-oh/5364?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart