CH6371380389 Chem Impex International, Inc.

Ethyleneglycolmonooctylether

  • MDL number MFCD00043070
  • Cas Number : 10020-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form Clear colorless liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ethyleneglycolmonooctylether/5368?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ethyleneglycolmonooctylether/5368?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart