CH6371380390 Chem Impex International, Inc.

Ethyleneglycolmonopentylether

  • MDL number MFCD00077908
  • Cas Number : 6196-58-3
  • Molecular Weight : 132.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8°C
Physical Form Clear, colorless liquid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ethyleneglycolmonopentylether/5369?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ethyleneglycolmonopentylether/5369?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart