CH6371380393 Chem Impex International, Inc.

Fmoc-S-sulfo-L-cysteine disodium

  • MDL number MFCD00237016
  • Cas Number : 163558-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Cys(SO3H)-OH disodium salt
Purity 99.8
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-sulfo-l-cysteine-disodium/5377?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-sulfo-l-cysteine-disodium/5377?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart