CH6371380394 Chem Impex International, Inc.

Na-Fmoc-D-arginine hydrochloride

  • MDL number MFCD01318732
  • Cas Number : 130752-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-Arg-OH HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-d-arginine-hydrochloride/5378?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-d-arginine-hydrochloride/5378?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart