CH6371380395 Chem Impex International, Inc.

Fmoc-D-aspartic acid b-benzyl ester

  • MDL number MFCD00273445
  • Cas Number : 150009-58-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-128
Synonym Fmoc-D-Asp(OBzl)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-aspartic-acid-beta-benzyl-ester/5379?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-aspartic-acid-beta-benzyl-ester/5379?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart