CH6371380399 Chem Impex International, Inc.

Fmoc-Ne-2,4-dinitrophenyl-D-lysine

  • MDL number MFCD01318743
  • Cas Number : 269061-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 145-151
Synonym Fmoc-D-Lys(Dnp)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form Yellow crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-sup-epsilon-sup-2-4-dinitrophenyl-d-lysine/5383?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-n-i-sup-epsilon-sup-2-4-dinitrophenyl-d-lysine/5383?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart