CH6371380400 Chem Impex International, Inc.

Na,e-Bis-Fmoc-D-lysine

  • MDL number MFCD00270743
  • Cas Number : 75932-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-144 �C
Synonym Fmoc-D-Lys(Fmoc)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-epsilon-sup-bis-fmoc-d-lysine/5384?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-epsilon-sup-bis-fmoc-d-lysine/5384?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart