CH6371380401 Chem Impex International, Inc.

Na-Fmoc-Ne-4-methoxyyltrityl-L-lysine

  • MDL number MFCD00270542
  • Cas Number : 159857-60-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-130 �C
Synonym Fmoc-L-Lys(Mmt)-OH
Purity 99.8
Storage temp. Store at 0-8
Physical Form White to pale brown solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-4-methoxyyltrityl-l-lysine/5385?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-4-methoxyyltrityl-l-lysine/5385?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart