CH6371380407 Chem Impex International, Inc.

Boc-1,5-diaminopentane HCl

  • MDL number MFCD02677949
  • Cas Number : 77835-31-5
  • Molecular Weight : 238.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-119 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-1-5-diaminopentane-font-size-2-face-symbol-middot-font-hcl/5399?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-1-5-diaminopentane-font-size-2-face-symbol-middot-font-hcl/5399?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart