CH6371380410 Chem Impex International, Inc.

Boc-D-His(1-Me)-OH

  • MDL number MFCD00237544
  • Cas Number : 200871-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-115 �C
Synonym Boc-D-His(Nim-Me)-OH
Purity 99
Storage temp. Store at =-4
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-his-1-me-oh/5408?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-his-1-me-oh/5408?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart