CH6371380411 Chem Impex International, Inc.

Boc-D-His(3-Me)-OH

  • MDL number MFCD00237545
  • Cas Number : 200871-84-7
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-184 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-his-3-me-oh/5410?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-his-3-me-oh/5410?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart