CH6371380414 Chem Impex International, Inc.

Phe-Gly-NH2�

  • MDL number MFCD00238115
  • Cas Number : 38678-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/phe-gly-nh-sub-2-sub-·hcl/5414?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/phe-gly-nh-sub-2-sub-·hcl/5414?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart