CH6371380416 Chem Impex International, Inc.

L-Glutamic acid gamma-methyl ester alpha-amide hydrochloride

  • MDL number MFCD02094379
  • Molecular Weight : 196.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190 - 210
Purity 98
Storage temp. Store at 0 - 8
Physical Form White powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/l-glutamic-acid-methyl-ester-a-amide-hydrochloride/5418?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamic-acid-methyl-ester-a-amide-hydrochloride/5418?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart