CH6371380418 Chem Impex International, Inc.

Fmoc-(2R,3S)-3-phenylisoserine

  • MDL number MFCD02259497
  • Cas Number : 252206-27-2
  • Molecular Weight : 403.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-199 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-2r-3-i-s-i-3-phenylisoserine/5420?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-2r-3-i-s-i-3-phenylisoserine/5420?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart