CH6371380422 Chem Impex International, Inc.

Benzyl-L-norleucine methyl ester hydrochloride

  • MDL number MFCD03093493
  • Cas Number : 1122017-35-9
  • Molecular Weight : 271.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Store at 0-8°C
Physical Form White powder with clumps
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzyl-l-norleucine-methyl-ester-hydrochloride/5424?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzyl-l-norleucine-methyl-ester-hydrochloride/5424?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart