CH6371380429 Chem Impex International, Inc.

4-(Fmoc-amino)-1-methyl-1H-imidazole-2-carboxylic acid

  • MDL number MFCD02259498
  • Cas Number : 252206-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-fmoc-amino-1-methyl-1h-imidazole-2-carboxylic-acid/5438?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-fmoc-amino-1-methyl-1h-imidazole-2-carboxylic-acid/5438?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart