CH6371380430 Chem Impex International, Inc.

Nalpha-Allyloxycarbonyl-Ndelta-Boc-D-ornithine dicyclohexyl ammonium salt

  • MDL number MFCD03093486
  • Cas Number : 947401-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-123 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-allyloxycarbonyl-i-n-i-sup-delta-sup-boc-d-ornithine-dicyclohexyl-ammonium-salt/5442?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-allyloxycarbonyl-i-n-i-sup-delta-sup-boc-d-ornithine-dicyclohexyl-ammonium-salt/5442?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart