CH6371380431 Chem Impex International, Inc.

Allyloxycarbonyl-D-phenylalanine dicyclohexyl ammonium salt

  • MDL number MFCD03093483
  • Cas Number : 152507-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-145
Synonym Aloc-D-Phe-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/allyloxycarbonyl-d-phenylalanine-dicyclohexyl-ammonium-salt/5443?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/allyloxycarbonyl-d-phenylalanine-dicyclohexyl-ammonium-salt/5443?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart