CH6371380433 Chem Impex International, Inc.

Na-Allyloxycarbonyl-Nd-Boc-L-ornithine dicyclohexyl ammonium salt

  • MDL number MFCD03093492
  • Cas Number : 171820-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-135
Synonym Aloc-L-Orn(Boc)-OH DCHA
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-allyloxycarbonyl-i-n-i-sup-delta-sup-boc-l-ornithine-dicyclohexyl-ammonium-salt/5446?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-allyloxycarbonyl-i-n-i-sup-delta-sup-boc-l-ornithine-dicyclohexyl-ammonium-salt/5446?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart