CH6371380437 Chem Impex International, Inc.

Boc-S-tert-butyl-L-cysteine dicyclohexylammonium salt

  • MDL number MFCD09039285
  • Cas Number : 60143-30-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-187 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-i-tert-i-butyl-l-cysteine-dicyclohexylammonium-salt/5450?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-i-tert-i-butyl-l-cysteine-dicyclohexylammonium-salt/5450?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart