CH6371380438 Chem Impex International, Inc.

Boc-trans-L-4-hydroxyproline benzyl ester

  • MDL number MFCD00076979
  • Cas Number : 89813-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 0-8 °C
Physical Form Yellowish oil
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-trans-i-l-4-hydroxyproline-benzyl-ester/5451?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-trans-i-l-4-hydroxyproline-benzyl-ester/5451?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart