CH6371380440 Chem Impex International, Inc.

Nalpha-Boc-L-lysine 7-amido-4-methylcoumarin acetate salt

  • MDL number MFCD03093404

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-l-lysine-7-amido-4-methylcoumarin/5453?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-l-lysine-7-amido-4-methylcoumarin/5453?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart