CH6371380441 Chem Impex International, Inc.

Na-Boc-Ne-4-nitro-Z-L-lysine

  • MDL number MFCD03093494
  • Cas Number : 22601-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-113 �C
Synonym Boc-L-Lys(4-NO2-Z)-OH
Purity 98
Storage temp. Store at =-4
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-4-nitro-z-l-lysine/5454?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-boc-i-n-i-sup-e-sup-4-nitro-z-l-lysine/5454?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart