CH6371380443 Chem Impex International, Inc.

Boc-b-(2-thienyl)-DL-alanine

  • MDL number MFCD00237537

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-117
Synonym Boc-3-DL-Ala(2-thienyl)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-beta-2-thienyl-dl-alanine/5456?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-beta-2-thienyl-dl-alanine/5456?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart