CH6371380446 Chem Impex International, Inc.

Fmoc-S-4-methyltrityl-L-cysteine

  • MDL number MFCD02259492
  • Cas Number : 269067-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-116
Synonym Fmoc-L-Cys(Mtt)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-4-methyltrityl-l-cysteine/5467?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-4-methyltrityl-l-cysteine/5467?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart