CH6371380447 Chem Impex International, Inc.

Fmoc-S-tert-butoxycarbonylmethyl-L-cysteine

  • MDL number MFCD02094571
  • Cas Number : 269730-62-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Cys(t-butoxycarbonylmethyl)-OH
Purity 97
Storage temp. Store at 0-8
Physical Form Syrup to White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-i-tert-i-butoxycarbonylmethyl-l-cysteine/5468?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-s-i-i-tert-i-butoxycarbonylmethyl-l-cysteine/5468?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart