CH6371380449 Chem Impex International, Inc.

Fmoc-a-methyl-D-phenylalanine

  • MDL number MFCD02259493
  • Cas Number : 152436-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-alpha-Me-D-Phe-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-a-methyl-d-phenylalanine/5471?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-a-methyl-d-phenylalanine/5471?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart