CH6371380450 Chem Impex International, Inc.

Nalpha-Fmoc-Nim-methyl-L-histidine

  • MDL number MFCD02259495
  • Cas Number : 202920-22-7
  • Molecular Weight : 391.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135 - 136 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-im-sup-methyl-l-histidine/5472?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-im-sup-methyl-l-histidine/5472?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart