CH6371380456 Chem Impex International, Inc.

Formyl-D-phenylglycine

  • MDL number MFCD03093479
  • Cas Number : 10419-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at =-4°C
Physical Form White solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/formyl-d-phenylglycine/5480?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/formyl-d-phenylglycine/5480?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart