CH6371380457 Chem Impex International, Inc.

D-3,5-Dibromotyrosine

  • MDL number MFCD01076162
  • Cas Number : 50299-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 236-242 �C
Synonym 3,5-Dibromo-D-Tyr-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-3-5-dibromotyrosine/5482?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-3-5-dibromotyrosine/5482?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart