CH6371380458 Chem Impex International, Inc.

L-a-Aminobutyric acid amide

  • MDL number MFCD00136552
  • Cas Number : 143164-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-226 �C
Synonym L-Abu-NH2
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-a-aminobutyric-acid-amide/5483?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-a-aminobutyric-acid-amide/5483?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart