CH6371380459 Chem Impex International, Inc.

D-2-Cyclohexylglycine trifluoroacetate

  • MDL number MFCD02262223

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Chg-OH TFA
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-2-cyclohexylglycine-trifluoroacetate/5484?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-2-cyclohexylglycine-trifluoroacetate/5484?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart