CH6371380462 Chem Impex International, Inc.

Ne-Boc-D-lysine allyl ester hydrochloride

  • MDL number MFCD01318762
  • Cas Number : 218962-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-116
Synonym D-Lys(Boc)-OAll HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-d-lysine-allyl-ester-hydrochloride/5490?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-epsilon-sup-boc-d-lysine-allyl-ester-hydrochloride/5490?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart