CH6371380463 Chem Impex International, Inc.

D-Phenylglycine amide hydrochloride

  • MDL number MFCD01863288
  • Cas Number : 63291-39-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Phg-NH2 HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-phenylglycine-amide-hydrochloride/5491?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-phenylglycine-amide-hydrochloride/5491?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart