CH6371380466 Chem Impex International, Inc.

D-Tryptophan benzyl ester hydrochloride

  • MDL number MFCD00237293
  • Cas Number : 22839-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203-207 �C
Synonym D-Trp-OBzl HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/d-tryptophan-benzyl-ester-hydrochloride/5494?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/d-tryptophan-benzyl-ester-hydrochloride/5494?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart