CH6371380467 Chem Impex International, Inc.

L-Glutamic acid a-tert-butyl ester hydrochloride

  • MDL number MFCD02684516
  • Cas Number : 144313-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Glu-OtBu HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-glutamic-acid-a-i-tert-i-butyl-ester-hydrochloride/5495?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamic-acid-a-i-tert-i-butyl-ester-hydrochloride/5495?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart