CH6371380469 Chem Impex International, Inc.

L-Glutamic acid g-methyl ester a-tert-butyl ester hydrochloride

  • MDL number MFCD02094380
  • Cas Number : 34582-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129-133
Synonym L-Glu(OMe)-OtBu HCl
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-glutamic-acid-gamma-methyl-ester-alpha-i-tert-i-butyl-ester-hydrochloride/5497?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamic-acid-gamma-methyl-ester-alpha-i-tert-i-butyl-ester-hydrochloride/5497?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart