CH6371380470 Chem Impex International, Inc.

L-Glutamic acid g-tert-butyl ester a-allyl ester hydrochloride

  • MDL number MFCD03093487
  • Cas Number : 1214283-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Glu(OtBu)-OAll HCl
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-glutamic-acid-gamma-i-tert-i-butyl-ester-alpha-allyl-ester-hydrochloride/5498?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamic-acid-gamma-i-tert-i-butyl-ester-alpha-allyl-ester-hydrochloride/5498?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart