CH6371380471 Chem Impex International, Inc.

Ne-Nicotinyl-L-lysine hydrochloride

  • MDL number MFCD03093488
  • Cas Number : 158276-23-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Lys(nicotinoyl)-OH HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-e-sup-nicotinyl-l-lysine-hydrochloride/5499?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-e-sup-nicotinyl-l-lysine-hydrochloride/5499?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart