CH6371380472 Chem Impex International, Inc.

4-Fluoro-D-Phenylalanine methyl ester hydrochloride

  • MDL number MFCD02259532
  • Cas Number : 176896-72-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169-175
Synonym 4-Fluoro-D-Phe-OMe HCl
Purity 99
Storage temp. Store at 0-8
Physical Form Snow white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/4-fluoro-d-phenylalanine-methyl-ester-hydrochloride/5500?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/4-fluoro-d-phenylalanine-methyl-ester-hydrochloride/5500?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart