CH6371380476 Chem Impex International, Inc.

b-Cyclohexyl-D-alanine hydrochloride

  • MDL number MFCD00190860
  • Cas Number : 151899-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Cha-OH HCl
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ß-cyclohexyl-d-alanine-hydrochloride/5504?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ß-cyclohexyl-d-alanine-hydrochloride/5504?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart