CH6371380489 Chem Impex International, Inc.

Z-L-glutamic acid g-N-hydroxysuccinimide ester a-benzyl ester

  • MDL number MFCD00237349
  • Cas Number : 34897-67-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 101-107
Synonym Z-L-Glu(OSu)-OBzl
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-g-i-n-i-hydroxysuccinimide-ester-a-benzyl-ester/5524?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-glutamic-acid-g-i-n-i-hydroxysuccinimide-ester-a-benzyl-ester/5524?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart