CH6371380491 Chem Impex International, Inc.

Z-O-trityl-L-serine

  • MDL number MFCD02684513
  • Cas Number : 1330286-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-186
Synonym Z-L-Ser(Trt)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-o-i-trityl-l-serine/5527?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-o-i-trityl-l-serine/5527?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart